Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Stoppers for Conical Ground Joints | Avantor
Stoppers for Conical Ground Joints
29/32, Mit Abtropfspitze
Katalognr. 217-9110
Anbieter:   Lenz Laborglas
Stoppers for Conical Ground Joints
Katalognr. 217-9110
Anbieter:  Lenz Laborglas
Stoppers for Conical Ground Joints
Stoppers for Conical Ground Joints
29/32, Mit Abtropfspitze
Preis
SFr. 9.40
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
These standard taper hollow stoppers are made from borosilicate glass 3.3 and have a hexagonal head.
  • Excellent chemical resistance
  • High thermal resistance, minimal heat expansion results in high resistance to temperature changes
  • Hexagonal head allows easy release and removal of the stopper
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
 : DIN 12252
 : Available with flat or pointed bottom.
Documents
Technische Daten
  • NS:
    29/32
  • Typ:
    Mit Abtropfspitze
  • Seq:
    07
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.