Stoppers for Conical Ground Joints
29/32, Mit Abtropfspitze
Katalognr. 217-9110
Anbieter: Lenz Laborglas
Direkt zu
Product Details & Documents
These standard taper hollow stoppers are made from borosilicate glass 3.3 and have a hexagonal head.
- Excellent chemical resistance
- High thermal resistance, minimal heat expansion results in high resistance to temperature changes
- Hexagonal head allows easy release and removal of the stopper
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
: DIN 12252
: Available with flat or pointed bottom.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Technische Daten
- NS:29/32
- Typ:Mit Abtropfspitze
- Seq:07