Evaporating dishes, high shape
250 ml, 50 mm, 85 mm
Katalognr. BOCH8561
Anbieter: BOCHEM
Direkt zu
Product Details & Documents
Evaporating dish, stainless steel, with contact bottom for heating plates and magnetic stirrers.
- Very good temperature resistance
- Good chemical resistance
- Easy to clean
Technische Daten
- Ext.Ø:85 mm
- Kapazität:250 ml
- Höhe:50 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



