Ethylendiamin 99+%
Glasflasche, 250 mL
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: 1,2-Ethanediamine, 1,2-Diaminoethane
- CAS Nummer:107-15-3
- Formel:C₂H₈N₂
- MDL-Nummer:MFCD00008204
- Molekulargewicht:60.1 g/mol
- Merck-Index:13,03829
- Siedepunkt:117…118 °C (760 mmHg)
- Schmelzender Platingehalt:11 °C
- Dichte:0,898 g/cm³
- Blitz Pt:38 °C
- Lagertemperatur:Raumtemperatur
- EINECS:203-468-6
- UN:1604
- ADR:8,II
Dokumente
ACRO22042
Sicherheitsdatenblatt (SDS)
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4555 to 1.4575 (20°C, 589 nm)
- Water:≤0.5 % (Coulometric)
- GC:≥99.0 %
- Color scale:≤30 APHA
Technische Daten
- Packung:Glasflasche
- VE:250 mL
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



