Kupfer(II)oxid 97%
Glasflasche für Feststoffe, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Cupric oxide, Copper oxide, Copper monoxide granulated
- CAS Number:1317-38-0
- Formula:CuO
- MDL Number:MFCD00010979
- Molecular Weight:79.55 g/mol
- Merck Index:13,02674
- Boiling Pt:2000 °C (1013 hPa)
- Melting Pt:1326 °C
- Density:4,9 g/cm³ (20 °C)
- Storage Temperature:Raumtemperatur
- EINECS:215-269-1
- UN:3077
- ADR:9,III
Documents
ACRO19744
SDS
Specifications
Specification Test Results
- Appearance:Black Powder
- Titration Iodometric:≥96.0 %
Specifications
- Packung:Glasflasche für Feststoffe
- VE:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



