Natriumalginat
Kst.-Flasche, 250 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: Alginic acid sodium salt, Algin
- CAS Nummer:9005-38-3
- MDL-Nummer:MFCD00081310
- Lagertemperatur:Raumtemperatur
Documents
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:Off-white to yellow-brown Powder
- Infrared spectrum:Conforms
- Loss on drying:≤15.0 % (100 mg, 105°C, 4 hrs)
- Sulfated ash:30.0 % to 36.0 % (on dried substance)
- Viscosity:350 to 550 mPa.s (1 % at 20°C) (Brookfield LV)
- pH:5.5 to 8.0 (1 % at 20°C)
- Heavy metals (as Pb):≤20 ppm
Technische Daten
- Packung:Kst.-Flasche
- VE:250 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



