Natriumalginat
Glasflasche für Feststoffe, 5 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: Alginic acid sodium salt, Algin
- CAS Nummer:9005-38-3
- MDL-Nummer:MFCD00081310
- Lagertemperatur:Raumtemperatur
Dokumente
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:Off-white to yellow-brown Powder
- Infrared spectrum:Conforms
- Loss on drying:≤15.0 % (100 mg, 105°C, 4 hrs)
- Sulfated ash:30.0 % to 36.0 % (on dried substance)
- Viscosity:350 to 550 mPa.s (1 % at 20°C) (Brookfield LV)
- pH:5.5 to 8.0 (1 % at 20°C)
- Heavy metals (as Pb):≤20 ppm
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:5 g
- Cat. No.:ACRO177770050
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



