D-Alanin 99+%
Glasflasche für Feststoffe, 5 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: (R)-(-)-Alanine, Alanine, D-α-Alanine, D(-)-Alanine, H-D-Ala-OH, α-Alanine
- CAS Nummer:338-69-2
- Formel:C₃H₇NO₂
- MDL-Nummer:MFCD00008077
- Molekulargewicht:89.09 g/mol
- Merck-Index:13,00203
- Siedepunkt:189 °C (1013 hPa)
- Schmelzender Platingehalt:278…282 °C (Zers.)
- Schmelzender Platingehalt:278…282 °C (Zersetzung)
- Lagertemperatur:Raumtemperatur
- EINECS:206-418-1
Documents
ACRO17208
SDS
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:White to off-white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤0.2 % (105°C, 3 hrs)
- Sulfated ash:≤0.2 %
- HPLC:≤1 % (L-content)
- UV transmittance:≥95 % (c=10, H2O, 430 nm) 1 cm cell
- Specific optical rotation:-13.5° to -15.5° (20°C, 589 nm) (c=10, 6 N HCl) (on dry substance)
- Titration with HClO4:99.0 to 101.0 % (On dry substance)
- Chloride (Cl):≤0.1 %
- Arsenic (As):≤2 ppm (As2O3)
- Heavy metals (as Pb):≤10 ppm
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:5 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



