Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Magnetic stirring bars, plain, PTFE

Magnetic stirring bars, plain, PTFE

12 mm, 4,5 mm
Catalog # COWI001.212
Supplier:   COWIE
Magnetic stirring bars, plain, PTFE
Catalog # COWI001.212
Supplier:  COWIE
Magnetic stirring bars, plain, PTFE
Magnetic stirring bars, plain, PTFE

12 mm, 4,5 mm

Price
SFr. 1.90
Log in for availability
1 * 1 ST
Product Details & Documents

Plain PTFE magnetic stirring bars have similar performance to cylindrical bars, but provide more turbulence at low speeds.

Specifications
  • Ø:
    4,5 mm
  • Länge:
    12 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.