Pipette tips, microvolume, Corning®
Runde Spitze, 0,5 - 10 µl, -, Transparent, Eppendorf, andere Ultra-Mikropipettoren, Lose; 10.000 Spitzen/Beutel
Anbieter: Corning
Direkt zu
Product Details & Documents
These microvolume tips provide accurate, reliable performance and are designed for use with major brand ultra-micropipettors.
- All racked tips are certified RNase-/DNase-free and non pyrogenic
- Exceptional durability: all tip racks stand up to the demands of use with multi-channel pipettors and withstand repeated autoclaving cycles
Dokumente
Zertifikate
Suche nach ChargennummerTechnische Daten
- Verpackt:Lose; 10.000 Spitzen/Beutel
- Steril:-
- Volumen:0,5 - 10 µl
- Spitzentyp:Runde Spitze
- Spitzenfarbe:Transparent
- Zur Nutzung mit:Eppendorf, andere Ultra-Mikropipettoren