96-well solid black and white polystyrene microplates, Corning®
Flat bottom, untreated, without lid, Schwarz, 75 - 200 µl, 360 µl, -, 25/bag
Anbieter: Corning
96-well solid black and white polystyrene microplates, Corning®
Katalognr. 734-1663
Anbieter: Corning
Direkt zu
Product Details & Documents
These 96-well solid black and white polystyrene microplates are designed to reduce well-to-well cross-talk.
White microplates enhance luminescent signals and have low background luminescence.
Black microplates have low background fluorescence and minimise light scattering.
Technische Daten
- Verpackt:25/bag
- Empf. Arbeitsvolumen:75 - 200 µl
- Steril:-
- Farbe:Schwarz
- Bezeichnung:Flat bottom, untreated, without lid
- Seq:18
- Wellvolumen:360 µl