AuftragsverfolgungKontakt
96-well solid black and white polystyrene microplates, Corning®
Flat bottom, untreated, without lid, Schwarz, 75 - 200 µl, 360 µl, -, 25/bag
Katalognr. 734-1663
Anbieter:   Corning
96-well solid black and white polystyrene microplates, Corning®
Katalognr. 734-1663
Anbieter:  Corning
96-well solid black and white polystyrene microplates, Corning®
96-well solid black and white polystyrene microplates, Corning®
Flat bottom, untreated, without lid, Schwarz, 75 - 200 µl, 360 µl, -, 25/bag
Preis
SFr. 633.00
Einloggen für Verfügbarkeit
1 * 100 ST
Product Details & Documents
These 96-well solid black and white polystyrene microplates are designed to reduce well-to-well cross-talk.
White microplates enhance luminescent signals and have low background luminescence.
Black microplates have low background fluorescence and minimise light scattering.
Technische Daten
  • Verpackt:
    25/bag
  • Empf. Arbeitsvolumen:
    75 - 200 µl
  • Steril:
    -
  • Farbe:
    Schwarz
  • Bezeichnung:
    Flat bottom, untreated, without lid
  • Seq:
    18
  • Wellvolumen:
    360 µl