2-Propanol 99.5+%
Glasflasche, 2,5 L
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
- CAS Nummer:67-63-0
- Formel:(CH₃)₂CHOH
- MDL-Nummer:MFCD00011674
- Molekulargewicht:60.1 g/mol
- Merck-Index:13,05228
- Siedepunkt:82 °C (1013 hPa)
- Schmelzender Platingehalt:–89,5 °C
- Dichte:0,785 g/cm³
- Blitz Pt:12 °C
- Lagertemperatur:Raumtemperatur
- EINECS:200-661-7
- UN:1219
- ADR:3,II
- ERREICHEN:01-2119457558-25
Documents
ACRO14932
SDS
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.2 % (K.F.)
- GC:≥99.50 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %
Technische Daten
- Packung:Glasflasche
- VE:2,5 L
- Cat. No.:ACRO149320025
- Umweltzertifizierung:Chem21 Solvent Selection Guide recommended after discussion
- Umweltfreundlichkeit:Unbedenklicher für Mensch und Umwelt
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



