2-Propanol 99.5+%
Glasflasche, 1 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
- CAS Number:67-63-0
- Formula:(CH₃)₂CHOH
- MDL Number:MFCD00011674
- Molecular Weight:60.1 g/mol
- Merck Index:13,05228
- Boiling Pt:82 °C (1013 hPa)
- Melting Pt:–89,5 °C
- Density:0,785 g/cm³
- Flash Pt:12 °C
- Storage Temperature:Raumtemperatur
- EINECS:200-661-7
- UN:1219
- ADR:3,II
- REACH:01-2119457558-25
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.2 % (K.F.)
- GC:≥99.50 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %
Specifications
- Packung:Glasflasche
- VE:1 L



