L-Threonin 98%
Glasflasche, 100 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid
- CAS Nummer:72-19-5
- Formel:C₄H₉NO₃
- MDL-Nummer:MFCD00064270
- Molekulargewicht:119.12 g/mol
- Schmelzender Platingehalt:255 °C (Zers.)
- Schmelzender Platingehalt:255 °C (Zersetzung)
- Lagertemperatur:Raumtemperatur
- EINECS:200-774-1
Documents
ACRO13893
SDS
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:White to off-white Crystalline powder or glistening flakes
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (1 g, 105°C, 3 hrs)
- Sulfated ash:≤0.1 %
- Solubility:(2.5 % in water) Clear colorless
- Specific optical rotation:-27.6° to -29.0° (20°C, 589 nm) (c=6, H2O)
- Titration with HClO4:97.5 to 102.5 % (On dried substance)
- Chloride (Cl):≤200 ppm
- Sulfate (SO4):≤300 ppm
- Heavy metals (as Pb):≤10 ppm
Technische Daten
- Packung:Glasflasche
- VE:100 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



