Natriumpyruvat 99+%
Kst.-Flasche, 100 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- CAS Nummer:113-24-6
- Formel:CH₃COCOONa
- MDL-Nummer:MFCD00002586
- Molekulargewicht:110.04 g/mol
- Schmelzender Platingehalt:>300 °C
- Dichte:1,718 g/cm³ (20 °C)
- Lagertemperatur:Kühlschrank
- EINECS:204-024-4
Documents
ACRO13215
SDS
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 %
- Heavy metals:≤10 ppm
- Solubility:(10 % in water) Clear colorless to light yellow
- Titration with HClO4:≥99.0 %
- Chloride (Cl):≤20 ppm
- Sulfate (SO4):≤20 ppm
- Arsenic (As):≤1 ppm
Technische Daten
- Packung:Kst.-Flasche
- VE:100 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



