Natriumpyruvat 99+%
Kst.-Flasche, 25 g
: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- :113-24-6
- :CH₃COCOONa
- :MFCD00002586
- :110.04 g/mol
- :>300 °C
- :1,718 g/cm³ (20 °C)
- :Kühlschrank
- :204-024-4
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 %
- Heavy metals:≤10 ppm
- Solubility:(10 % in water) Clear colorless to light yellow
- Titration with HClO4:≥99.0 %
- Chloride (Cl):≤20 ppm
- Sulfate (SO4):≤20 ppm
- Arsenic (As):≤1 ppm
- Packung:Kst.-Flasche
- VE:25 g



