Inosin 99%
Glasflasche, 100 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: 9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one
- CAS Nummer:58-63-9
- Formel:C₁₀H₁₂N₄O₅
- MDL-Nummer:MFCD00066770
- Molekulargewicht:268.23 g/mol
- Schmelzender Platingehalt:212…213 °C (Zers.)
- Schmelzender Platingehalt:212…213 °C (Zersetzung)
- Lagertemperatur:Raumtemperatur
- EINECS:200-390-4
Documents
ACRO12225
SDS
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 %
- Sulfated ash:≤0.1 %
- HPLC:≥98.5 %
- UV spectrophotometry:98 to 102 % (lambda max. 248.5nm, pH 7)
- UV transmittance:≥98 % (c=1, H2O, 430 nm, 1 cm cell)
- Ratio of Absorbances:1.65 to 1.77 (A250/A260)
- Titration with HClO4:≥98.5 % (On dry substance)
- Heavy metals (as Pb):≤10 ppm
Technische Daten
- Packung:Glasflasche
- VE:100 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



