Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Inosin 99%

Inosin 99%

Glasflasche, 10 g
Catalog # ACRO122250100
Inosin 99%
Catalog # ACRO122250100
CAS Number:  58-63-9
Inosin 99%
Inosin 99%

Glasflasche, 10 g

CAS Number:  58-63-9
Price
SFr. 76.40
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one
  • CAS Number:58-63-9
  • Formula:C₁₀H₁₂N₄O₅
  • MDL Number:MFCD00066770
  • Molecular Weight:268.23 g/mol
  • Melting Pt:212…213 °C (Zers.)
  • Melting Pt:212…213 °C (Zersetzung)
  • Storage Temperature:Raumtemperatur
  • EINECS:200-390-4

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 %
  • Sulfated ash:
    ≤0.1 %
  • HPLC:
    ≥98.5 %
  • UV spectrophotometry:
    98 to 102 % (lambda max. 248.5nm, pH 7)
  • UV transmittance:
    ≥98 % (c=1, H2O, 430 nm, 1 cm cell)
  • Ratio of Absorbances:
    1.65 to 1.77 (A250/A260)
  • Titration with HClO4:
    ≥98.5 % (On dry substance)
  • Heavy metals (as Pb):
    ≤10 ppm

Specifications

  • Packung:
    Glasflasche
  • VE:
    10 g

Specifications

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..