(4-Hydroxyphenyl)essigsäure 98%
Glasflasche für Feststoffe, 25 g
Supplier: Thermo Fisher Scientific
(4-Hydroxyphenyl)essigsäure 98%
Catalog # ACRO121710250
Supplier: Thermo Fisher Scientific
CAS Number: 156-38-7
Jump to:
Product Details & Documents
Synonyms: 4-Hydroxyphenylacetic Acid
- CAS Number:156-38-7
- Formula:(C₆H₄(OH))CH₂COOH
- MDL Number:MFCD00004347
- Molecular Weight:152.15 g/mol
- Boiling Pt:347 °C (1013 hPa)
- Melting Pt:147…152 °C
- Storage Temperature:Raumtemperatur
- EINECS:205-851-3
Specifications
Specification Test Results
- Appearance:White to cream to tan Crystalline powder
- Infrared spectrum:Conforms
- Melting point:147 to 152 °C
- Water:≤0.5 % (K.F.)
- TLC:≥97.5 %
- Titration with NaOH:97.5 to 102.5 %
Specifications
- Packung:Glasflasche für Feststoffe
- VE:25 g