Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
1,6-Diaminohexan 99.5+%

1,6-Diaminohexan 99.5+%

Kst.-Flasche, 100 g
Catalog # ACRO120641000
Irritant
Corrosive
1,6-Diaminohexan 99.5+%
Catalog # ACRO120641000
CAS Number:  124-09-4
1,6-Diaminohexan 99.5+%
1,6-Diaminohexan 99.5+%

Kst.-Flasche, 100 g

CAS Number:  124-09-4
Price
SFr. 36.30
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1,6-Hexanediamine, Hexamethylenediamine
  • CAS Number:124-09-4
  • Formula:C₆H₁₆N₂
  • MDL Number:MFCD00008243
  • Molecular Weight:116.21 g/mol
  • Merck Index:13,04713
  • Boiling Pt:204…205 °C (760 mmHg)
  • Melting Pt:38…41 °C
  • Density:0,66 g/cm³ (20 °C)
  • Flash Pt:81 °C
  • Storage Temperature:Raumtemperatur
  • EINECS:204-679-6
  • UN:2280
  • ADR:8,III
Specifications

Specification Test Results

  • Appearance:
    White Crystalline low melting mass (May discolor on storage)
  • Infrared spectrum:
    Conforms
  • Melting point:
    38 to 41 °C
  • Water:
    ≤0.05 % (K.F.)
  • GC:
    ≥99.50 %
  • Titration with HClO4:
    99.5 to 100.5 %

Specifications

  • Packung:
    Kst.-Flasche
  • VE:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.