Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
(±)-Epichlorhydrin 99%

(±)-Epichlorhydrin 99%

Glasflasche, 2,5 L
Catalog # ACRO117780025
Toxic
Health
Corrosive
Flammable
(±)-Epichlorhydrin 99%
Catalog # ACRO117780025
CAS Number:  106-89-8
(±)-Epichlorhydrin 99%
(±)-Epichlorhydrin 99%

Glasflasche, 2,5 L

CAS Number:  106-89-8
Price
SFr. 173.00
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  (±)-1-Chloro-2,3-epoxypropane, Epichlorohydrin, (±)-3-Chloro-1,2-epoxypropane, 1-Chloro-2,3-epoxypropane, Epichlorhydrin, (±)-Glycerol epichlorhydrin, 3-Chloro-1,2-epoxypropane
  • CAS Number:106-89-8
  • Formula:C₃H₅ClO
  • MDL Number:MFCD00005132
  • Molecular Weight:92.52 g/mol
  • Merck Index:13,03642
  • Boiling Pt:115…117 °C (760 mmHg)
  • Melting Pt:–57 °C
  • Density:1,18 g/cm³
  • Flash Pt:28 °C
  • Storage Temperature:Kühlschrank
  • EINECS:203-439-8
  • UN:2023
  • ADR:6.1,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4365 to 1.4385 (20°C, 589 nm)
  • Water:
    ≤0.02 % (K.F.)
  • GC:
    ≥98.5 %
  • Color scale:
    ≤15 APHA
  • Silicate and phosphate:
    ≤5 ppm (as SiO2)
  • Iron (Fe):
    ≤1 ppm
  • Heavy metals (as Pb):
    ≤1 ppm
  • Nitrogen compounds (as N):
    ≤0.001 %

Specifications

  • Packung:
    Glasflasche
  • VE:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.