1,4-Dimethoxybenzol 99+%
Glasflasche für Feststoffe, 500 g
Anbieter: Thermo Fisher Scientific
1,4-Dimethoxybenzol 99+%
Katalognr. ACRO115415000
Anbieter: Thermo Fisher Scientific
CAS Nummer: 150-78-7
Direkt zu
Product Details & Documents
Synonyme: p-Dimethoxybenzene, Hydroquinone dimethyl ether
- CAS Nummer:150-78-7
- Formel:C₈H₁₀O₂
- MDL-Nummer:MFCD00008401
- Molekulargewicht:138.17 g/mol
- Siedepunkt:212,6 °C (1013 hPa)
- Schmelzender Platingehalt:54…58 °C
- Dichte:1,05 g/cm³
- Blitz Pt:98 °C
- Lagertemperatur:Raumtemperatur
- EINECS:205-771-9
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:White to off-white to grey Crystals of flakes
- Infrared spectrum:Conforms
- Melting point:54 to 58 °C
- GC:≥99.0 %
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:500 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



