Bicin, Calbiochem®
Glasflasche für Feststoffe, 100 g
Katalognr. 391336-100
Anbieter: Merck Millipore (Calbiochem)
Bicin, Calbiochem®
Katalognr. 391336-100
Anbieter: Merck Millipore (Calbiochem)
CAS Nummer: 150-25-4
Direkt zu
Product Details & Documents
Synonyme: 2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine
- CAS Nummer:150-25-4
- Formel:C₆H₁₃NO₄
- MDL-Nummer:MFCD00004295
- Molekulargewicht:163.17 g/mol
- Schmelzender Platingehalt:185…192 °C
- Lagertemperatur:Raumtemperatur
- EINECS:205-755-1
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:100 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



