4-Phenylbenzaldehyd 99%
Glasflasche für Feststoffe, 5 g
Anbieter: Thermo Fisher Scientific
4-Phenylbenzaldehyd 99%
Katalognr. ACRO106260050
Anbieter: Thermo Fisher Scientific
CAS Nummer: 3218-36-8
Direkt zu
Product Details & Documents
Synonyme: Biphenyl-4-carbaldehyde, [1,1'-Biphenyl]-4-carbaldehyde, 4-Biphenylcarboxaldehyde, Biphenyl-4-carboxaldehyde, p-Biphenylylaldehyde, 4-Biphenylylaldehyde, 4-Biphenylcarbaldehyde
- CAS Nummer:3218-36-8
- Formel:C₁₃H₁₀O
- MDL-Nummer:MFCD00006947
- Molekulargewicht:182.22 g/mol
- Siedepunkt:184 °C (11 mmHg)
- Schmelzender Platingehalt:57…60 °C
- Lagertemperatur:Raumtemperatur
- EINECS:221-742-3
Documents
ACRO10626
SDS
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:White to light yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:57 to 60 °C
- GC:≥98.5 %
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:5 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



