Vanadium(V)oxid ≥99,6%
Glass bottle for solids, 500 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99.6% min CAS No.: 1314-62-1 Molecular Formula: V2O5 Formula Weight: 181.88 Physical Form: -10 Mesh Density: 3.357 Melting Point: 690° Boiling Point: 1750° dec. Merck Reference: 14,9921 UN No.: 2862 MDL No.: MFCD00011457
Synonyme: Vanadium pentoxide, di-Vanadium pentaoxide
- CAS Nummer:1314-62-1
- Formel:O₅V₂
- MDL-Nummer:MFCD00011457
- Molekulargewicht:181.88 g/mol
- Merck-Index:14,9921
- Siedepunkt:1750 °C (Zers.)
- Siedepunkt:1750 °C (Zersetzung)
- Schmelzender Platingehalt:690 °C
- Dichte:3,357
- Lagertemperatur:Raumtemperatur
- EINECS:215-239-8
- UN:2862
- ADR:6.1,III
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:500 g