Schwefelsäure 95% ACS
Glasflasche, 500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:7664-93-9
- Formula:H₂SO₄
- MDL Number:MFCD00064589
- Molecular Weight:98.08 g/mol
- Merck Index:14,08974
- Boiling Pt:∼335 °C (1013 hPa)
- Melting Pt:10 °C
- Density:1,83 g/cm³
- Storage Temperature:Raumtemperatur
- EINECS:231-639-5
- ADR:8,II
- :Density: 1.84 g/cm³ (20 °C)
- UN:1830
Documents
Specifications
Specification Test Results
- Appearance:Free from suspended or insoluble matter
- Color scale:≤10 APHA
- Residue after ignition:≤0.0005 %
- Titration with NaOH:95.0 to 98.0 %
- Chloride (Cl):≤0.2 ppm
- Nitrate (NO3):≤0.5 ppm
- Ammonium (NH4):≤2 ppm
- Arsenic (As):≤0.01 ppm
- Iron (Fe):≤0.2 ppm
- Mercury (Hg):≤5 ppb
- Heavy metals (as Pb):≤1 ppm
- KMnO4-reducing substance:≤2 ppm (as SO2)
Specifications
- Packung:Glasflasche
- VE:500 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



