Schwefelsäure 95% ACS
Glasflasche, 2,5 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:7664-93-9
- Formula:H₂SO₄
- MDL Number:MFCD00064589
- Molecular Weight:98.08 g/mol
- Merck Index:14,08974
- Boiling Pt:∼335 °C (1013 hPa)
- Melting Pt:10 °C
- Density:1,83 g/cm³
- Storage Temperature:Raumtemperatur
- EINECS:231-639-5
- ADR:8,II
- :Density: 1.84 g/cm³ (20 °C)
- UN:1830
Documents
Specifications
Specification Test Results
- Appearance:Free from suspended or insoluble matter
- Color scale:≤10 APHA
- Residue after ignition:≤0.0005 %
- Titration with NaOH:95.0 to 98.0 %
- Chloride (Cl):≤0.2 ppm
- Nitrate (NO3):≤0.5 ppm
- Ammonium (NH4):≤2 ppm
- Arsenic (As):≤0.01 ppm
- Iron (Fe):≤0.2 ppm
- Mercury (Hg):≤5 ppb
- Heavy metals (as Pb):≤1 ppm
- KMnO4-reducing substance:≤2 ppm (as SO2)
Specifications
- Packung:Glasflasche
- VE:2,5 L



