Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Morpholin 99+% ACS

Morpholin 99+% ACS

Glasflasche für Feststoffe, 5 g
Catalog # ACRO415160050
Toxic
Corrosive
Flammable
Morpholin 99+% ACS
Catalog # ACRO415160050
CAS Number:  110-91-8
Morpholin 99+% ACS
Morpholin 99+% ACS

Glasflasche für Feststoffe, 5 g

CAS Number:  110-91-8
Price
SFr. 26.10
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:110-91-8
  • Formula:C₄H₉NO
  • MDL Number:MFCD00005972
  • Molecular Weight:87.12 g/mol
  • Merck Index:12,06362
  • Boiling Pt:128,9 °C (1013 hPa)
  • Melting Pt:–5 °C
  • Density:0,99 g/cm³
  • Flash Pt:32 °C
  • Storage Temperature:Raumtemperatur
  • EINECS:203-815-1
  • UN:2054
  • ADR:8,I

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4530 to 1.4550 (20°C, 589 nm)
  • Boiling point:
    126.0 to 130.0°C
  • Color scale:
    ≤15 APHA
  • Titration with HCl:
    ≥99.0 %

Specifications

  • Packung:
    Glasflasche für Feststoffe
  • VE:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.