Methyldimethoxyacetat 98%
Glasflasche, 25 g
Anbieter: Thermo Fisher Scientific
Methyldimethoxyacetat 98%
Katalognr. ACRO414180250
Anbieter: Thermo Fisher Scientific
CAS Nummer: 89-91-8
Direkt zu
Product Details & Documents
Synonyme: Methyl 2,2-dimethoxyacetate
- CAS Nummer:89-91-8
- Formel:C₅H₁₀O₄
- MDL-Nummer:MFCD00008484
- Molekulargewicht:134.13 g/mol
- Siedepunkt:160…163 °C (760 mmHg)
- Dichte:1,09 g/cm³
- Blitz Pt:63 °C
- Lagertemperatur:Kühlschrank
- EINECS:201-950-0
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4040 to 1.4070 (20°C, 589 nm)
- GC:≥97.5 %
Technische Daten
- Packung:Glasflasche
- VE:25 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



