Lithiumchlorid 99+% ACS
Glasflasche für Feststoffe, 5 g
Supplier: Thermo Fisher Scientific
Lithiumchlorid 99+% ACS
Catalog # ACRO413270050
Supplier: Thermo Fisher Scientific
CAS Number: 7447-41-8
Jump to:
Product Details & Documents
- CAS Number:7447-41-8
- Formula:LiCl
- MDL Number:MFCD00011078
- Molecular Weight:42.39 g/mol
- Merck Index:13,05550
- Boiling Pt:1382 °C (760 mmHg)
- Melting Pt:605 °C
- Density:2,06 g/cm³
- Storage Temperature:Raumtemperatur
- EINECS:231-212-3
Documents
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Titration Argentometric:≥99 %
- Loss on drying:≤1.0 % (1 g, 105°C, 2 hrs)
- Sulfate (SO4):≤100 ppm
- Barium (Ba):≤30 ppm
- Calcium (Ca):≤100 ppm
- Iron (Fe):≤10 ppm
- Potassium (K):≤100 ppm
- Sodium (Na):≤2000 ppm
- Heavy metals (ICP-OES):≤20 ppm
- Titrable base:≤0.008 meq/g
- Insoluble matter:≤0.01 %
Specifications
- Packung:Glasflasche für Feststoffe
- VE:5 g



