Dosen mit Deckel
2500 ml, 150 mm, 150 mm
Katalognr. 216-0039
Anbieter: BOCHEM
Direkt zu
Product Details & Documents
Jar made of stainless steel, with knob lid.
- Very good temperature resistance
- Good chemical resistance
- Easy to clean
Technische Daten
- Ext.Ø:150 mm
- Kapazität:2500 ml
- Höhe:150 mm
- Seq:05