Dosen mit Deckel
400 ml, 85 mm, 85 mm
Katalognr. BOCH8584
Anbieter: BOCHEM
Direkt zu
Product Details & Documents
Jar made of stainless steel, with knob lid.
- Very good temperature resistance
- Good chemical resistance
- Easy to clean
Technische Daten
- Ext.Ø:85 mm
- Kapazität:400 ml
- Höhe:85 mm
- Seq:02