Spoon spatulas
230 mm, 25×12 mm
Katalognr. BOCH3722
Anbieter: BOCHEM
Direkt zu
Product Details & Documents
PTFE coated double spatulas with one spoon end and one flat end.
Technische Daten
- Länge:230 mm
- Löffelmaße:25×12 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



