Spoon spatulas
180 mm, 25×8 mm
Katalognr. BOCH3720
Anbieter: BOCHEM
Direkt zu
Product Details & Documents
PTFE coated double spatulas with one spoon end and one flat end.
Technische Daten
- Länge:180 mm
- Löffelmaße:25×8 mm
PTFE coated double spatulas with one spoon end and one flat end.