Actinomycin D 93%
Glasflasche, 25 mg
Red crystalline powder
- :50-76-0
- :C₆₂H₈₆N₁₂O₁₆
- :MFCD00005033
- :1255.5 g/mol
- :245…252 °C
- :Gefrierschrank
- :3462
- :6.1,II
- Appearance:Red Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥91 %
- Packung:Glasflasche
- VE:25 mg



