Actinomycin D 93%
Glasflasche, 5 mg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Red crystalline powder
- CAS Number:50-76-0
- Formula:C₆₂H₈₆N₁₂O₁₆
- MDL Number:MFCD00005033
- Molecular Weight:1255.5 g/mol
- Melting Pt:245…252 °C
- Storage Temperature:Gefrierschrank
- UN:3462
- ADR:6.1,II
Documents
ACRO29494
SDS
Specifications
Specification Test Results
- Appearance:Red Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥91 %
Specifications
- Packung:Glasflasche
- VE:5 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



