Triethylammoniumchlorid 99+%
Glasflasche für Feststoffe, 1 kg
Anbieter: Thermo Fisher Scientific
Triethylammoniumchlorid 99+%
Katalognr. ACRO270670010
Anbieter: Thermo Fisher Scientific
CAS Nummer: 554-68-7
Direkt zu
Product Details & Documents
Synonyme: Triethylamine hydrochloride
- CAS Nummer:554-68-7
- Formel:C₆H₁₆ClN
- MDL-Nummer:MFCD00012500
- Molekulargewicht:137.65 g/mol
- Schmelzender Platingehalt:254…260 °C
- Dichte:0,7637 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:209-067-2
Documents
ACRO27067
SDS
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:254 to 260 °C
- Loss on drying:≤1.0 %
- Solubility:(10 % in water) Clear colorless
- pH:4.5 to 6.5 (1 % w/v solution)
- Titration with HClO4:99.0 to 101.0 % (On anhydrous substance)
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:1 kg
- Cat. No.:ACRO270670010
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



