Rosolsäure (Aurin)
Glasflasche, 25 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: 4-(Bis(4-hydroxyphenyl)methylene)cyclohexa-2,5-dien-1-one
- CAS Nummer:603-45-2
- Formel:C₁₉H₁₄O₃
- MDL-Nummer:MFCD00001624
- Molekulargewicht:290.32 g/mol
- Lagertemperatur:Raumtemperatur
- EINECS:210-041-8
Documents
ACRO22983
SDS
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:Red-brown to brown Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤10 % (110°C, 1 h)
- UV Lambda max:480 to 486 nm (in ethanol + 2ml 1N HCl)
- Visual transition interval:from pH 6.2 (yellow) to pH 8.2 (red-pink)
Technische Daten
- Packung:Glasflasche
- VE:25 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



