Stopcocks for burettes, lateral
Büretten, Inhalt 25 ml
HIRS8901170
Lateral burette stopcock, made of borosilicate glass, with glass key, clear glass.
- Excellent chemical resistance
- Excellent temperature resistance
- Für:Büretten, Inhalt 25 ml
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.