Stopcocks for burettes, lateral
Büretten, Inhalt 2 - 10 ml
Katalognr. HIRS8901160
Anbieter: HIRSCHMANN
Direkt zu
Product Details & Documents
Lateral burette stopcock, made of borosilicate glass, with glass key, clear glass.
- Excellent chemical resistance
- Excellent temperature resistance
Technische Daten
- Für:Büretten, Inhalt 2 - 10 ml
- Seq:01