Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Stopcocks for burettes, lateral | Avantor
Stopcocks for burettes, lateral
Büretten, Inhalt 2 - 10 ml
Katalognr. HIRS8901160
Anbieter:   HIRSCHMANN
Stopcocks for burettes, lateral
Katalognr. HIRS8901160
Anbieter:  HIRSCHMANN
Stopcocks for burettes, lateral
Stopcocks for burettes, lateral
Büretten, Inhalt 2 - 10 ml
Preis
SFr. 41.30
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Lateral burette stopcock, made of borosilicate glass, with glass key, clear glass.
  • Excellent chemical resistance
  • Excellent temperature resistance
Technische Daten
  • Für:
    Büretten, Inhalt 2 - 10 ml
  • Seq:
    01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.