Messzylinder
1000 ml, 10 ml, 470 mm, 7 ± ml
612-3034
DURAN® Borosilikatglas, hohe Form, Klasse B.
- Calibrated to contain 'ln'
- Hexagonal glass base, with spout
- Amber stain graduations
* In Ergänzung zu DIN.
: DIN EN ISO 4788
- Kapazität:1000 ml
- Höhe:470 mm
- Seq:08
- Teilung:10 ml
- Toleranz:7 ± ml
- Cat. No.:612-3034