Messzylinder
500 ml, 5 ml, 390 mm, 3,75 ± ml
Katalognr. 612-3033
Anbieter: HIRSCHMANN
Direkt zu
Product Details & Documents
DURAN® Borosilikatglas, hohe Form, Klasse B.
- Calibrated to contain 'ln'
- Hexagonal glass base, with spout
- Amber stain graduations
* In Ergänzung zu DIN.
: DIN EN ISO 4788
Technische Daten
- Kapazität:500 ml
- Höhe:390 mm
- Seq:07
- Teilung:5 ml
- Toleranz:3,75 ± ml