Electron capture detectors (ECD)
Fused silica liner for micro ECD MUG adapter
Catalog # HEWLG2397-20540
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
The micro ECD has a detection zone volume 10 times smaller than any other ECD with replicable inlet liner to decreasing column contamination of the cell.
- Replaceable liner
The replaceable liner serves as a physical stop for the column, ensuring reproducible column installation and decreasing column contamination of the cell.
Specifications
- Bezeichnung:Fused silica liner for micro ECD MUG adapter
- Seq:05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.