AuftragsverfolgungKontakt
Electron capture detectors (ECD)
Electron capture detectors (ECD)
Fused silica liner for micro ECD MUG adapter
Catalog # HEWLG2397-20540
Electron capture detectors (ECD)
Catalog # HEWLG2397-20540
Electron capture detectors (ECD)
Electron capture detectors (ECD)
Fused silica liner for micro ECD MUG adapter
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
The micro ECD has a detection zone volume 10 times smaller than any other ECD with replicable inlet liner to decreasing column contamination of the cell.
  • Replaceable liner
The replaceable liner serves as a physical stop for the column, ensuring reproducible column installation and decreasing column contamination of the cell.
Specifications
  • Bezeichnung:
    Fused silica liner for micro ECD MUG adapter
  • Seq:
    05