Electron capture detectors (ECD)
ECD make-up gas adapter
N° de catalogue HEWLG1533-80565
Fournisseur: AGILENT HEWLETT PACKARD
Electron capture detectors (ECD)
N° de catalogue HEWLG1533-80565
Fournisseur: AGILENT HEWLETT PACKARD
Aller à
Product Details & Documents
The micro ECD has a detection zone volume 10 times smaller than any other ECD with replicable inlet liner to decreasing column contamination of the cell.
- Replaceable liner
The replaceable liner serves as a physical stop for the column, ensuring reproducible column installation and decreasing column contamination of the cell.
Spécifications
- Bezeichnung:ECD make-up gas adapter
- Seq:03