AuftragsverfolgungKontakt
Electron capture detectors (ECD)
Electron capture detectors (ECD)
ECD make-up gas adapter
N° de catalogue HEWLG1533-80565
Fournisseur:   AGILENT HEWLETT PACKARD
Electron capture detectors (ECD)
N° de catalogue HEWLG1533-80565
Electron capture detectors (ECD)
Electron capture detectors (ECD)
ECD make-up gas adapter
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
The micro ECD has a detection zone volume 10 times smaller than any other ECD with replicable inlet liner to decreasing column contamination of the cell.
  • Replaceable liner
The replaceable liner serves as a physical stop for the column, ensuring reproducible column installation and decreasing column contamination of the cell.
Spécifications
  • Bezeichnung:
    ECD make-up gas adapter
  • Seq:
    03