Inlet liners
Splitless, straight liner, deactivated, quartz, 250 μl, 2 mm
Katalognr. HEWL5183-4704
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Technische Daten
- Ø int.:2 mm
- Bezeichnung:Splitless, straight liner, deactivated, quartz, 250 μl
- Seq:41
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.