Inlet liners
PTV liner, single baffle, glass wool, deactivated, 180 µl, 2 mm
Katalognr. HEWL5183-2038
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Technische Daten
- Ø int.:2 mm
- Bezeichnung:PTV liner, single baffle, glass wool, deactivated, 180 µl
- Seq:60