Capillary column splitter kits
Fixed inlet splitter kit, 0,53 mm
Katalognr. HEWL5181-3392
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
These splitters are used for splitting an injected sample from a single injector into two columns and on to two detectors.
The dimensions of both the columns, length and internal diameter (ID), should be the same so that the vaporized sample will get spilt into almost equal amount on to two columns.
Lieferinformationen: Kit which comes with transfer line, splitter union and necessary ferrules.
Technische Daten
- Ø int.:0,53 mm
- Bezeichnung:Fixed inlet splitter kit
- Seq:03