Capillary column ferrules for gas chromatography
85% vespel, 15% graphite ferrules, short, ferrule ID 0,4 mm, column ID 0,1 to 0,25 mm
Katalognr. HEWL5181-3323
Anbieter: AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalognr. HEWL5181-3323
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
Technische Daten
- Bezeichnung:85% vespel, 15% graphite ferrules, short, ferrule ID 0,4 mm, column ID 0,1 to 0,25 mm
- Seq:08