Capillary column ferrules for gas chromatography
High performance ferrules, 100% vespel, short, ferrule ID 0,4 mm, column ID 0,1, 0,2, 0,25 mm
Katalognr. HEWL5181-3322
Anbieter: AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalognr. HEWL5181-3322
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
Documents
HEWL5062-3514_TDS_EN
Datasheet
Technische Daten
- Bezeichnung:High performance ferrules, 100% vespel, short, ferrule ID 0,4 mm, column ID 0,1, 0,2, 0,25 mm
- Seq:15