Capillary column ferrules for gas chromatography
General purpose graphite ferrules, short, ferrule ID 0,5 mm, column ID 0,1, 0,2, 0,25, 0,32 mm
Katalognr. HEWL5080-8853
Anbieter: AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalognr. HEWL5080-8853
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
Documents
HEWL5062-3514_TDS_EN
Datasheet
Technische Daten
- Bezeichnung:General purpose graphite ferrules, short, ferrule ID 0,5 mm, column ID 0,1, 0,2, 0,25, 0,32 mm
- Seq:04