AuftragsverfolgungKontakt
Capillary column ferrules for gas chromatography
Preconditioned 85% vespel, 15% graphite ferrules, long, ferrule ID 0,8 mm, column ID 0,53 mm
  HEWL5062-3538
Capillary column ferrules for gas chromatography
  HEWL5062-3538
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
Preconditioned 85% vespel, 15% graphite ferrules, long, ferrule ID 0,8 mm, column ID 0,53 mm
1 * 10 ST
  • Bezeichnung:
    Preconditioned 85% vespel, 15% graphite ferrules, long, ferrule ID 0,8 mm, column ID 0,53 mm
  • Seq:
    12