Capillary column ferrules for gas chromatography
Preconditioned 85% vespel, 15% graphite ferrules, long, ferrule ID 0,8 mm, column ID 0,53 mm
HEWL5062-3538
- Bezeichnung:Preconditioned 85% vespel, 15% graphite ferrules, long, ferrule ID 0,8 mm, column ID 0,53 mm
- Seq:12